2-Isobutyl-3-methoxypyrazine, 98%, Thermo Scientific Chemicals
2-Isobutyl-3-methoxypyrazine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Isobutyl-3-methoxypyrazine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A13680.03
also known as A13680-03
Price (USD)/ Each
57.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
57.00
Add to cart
2-Isobutyl-3-methoxypyrazine, 98%, Thermo Scientific Chemicals
Catalog numberA13680.03
Price (USD)/ Each
57.00
-
Add to cart
Chemical Identifiers
CAS24683-00-9
IUPAC Name2-methoxy-3-(2-methylpropyl)pyrazine
Molecular FormulaC9H14N2O
InChI KeyUXFSPRAGHGMRSQ-UHFFFAOYSA-N
SMILESCOC1=NC=CN=C1CC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.4895-1.4935 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only