Thermo Scientific Chemicals

Diethyl aminomalonate hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A13681.30
250 g, Each
Thermo Scientific Chemicals

Diethyl aminomalonate hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A13681.30
250 g, Each
Quantity
Catalog number: A13681.30
also known as A13681-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13433-00-6
IUPAC Name
hydrogen 1,3-diethyl 2-aminopropanedioate chloride
Molecular Formula
C7H14ClNO4
InChI Key
GLFVNTDRBTZJIY-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CCOC(=O)C(N)C(=O)OCC
Appearance (Color)
White
Form
Powder
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%

Description

Diethyl aminomalonate hydrochloride is used as a pharmaceutical intermediate. Diethyl aminomalonate with formaldehyde generates an azomethine ylide which can undergo cycloadditions with 1,2-dipolarophiles to form pyrrolidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl aminomalonate hydrochloride is used as a pharmaceutical intermediate. Diethyl aminomalonate with formaldehyde generates an azomethine ylide which can undergo cycloadditions with 1,2-dipolarophiles to form pyrrolidines.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is hygroscopic in nature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text