Thermo Scientific Chemicals

2,4'-Dibromoacetophenone, 98+%, Thermo Scientific Chemicals

Catalog number: A13687.22
100 g, Each
Thermo Scientific Chemicals

2,4'-Dibromoacetophenone, 98+%, Thermo Scientific Chemicals

Catalog number: A13687.22
100 g, Each
Quantity
Catalog number: A13687.22
also known as A13687-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-73-0
IUPAC Name
2-bromo-1-(4-bromophenyl)ethan-1-one
Molecular Formula
C8H6Br2O
InChI Key
FKJSFKCZZIXQIP-UHFFFAOYSA-N
SMILES
BrCC(=O)C1=CC=C(Br)C=C1
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
108.0-114.0?C
Appearance (Color)
White to pale cream

Description

2,4'-Dibromoacetophenone is used as a reagent for analysis of fatty acids by HPLC as 4-bromophenacyl esters, for use in the protection of phenols and carboxylic acids. It undergoes condensation reactions with aldehydes in the presence of SnCl2 or SmI3 to afford α,β-unsaturated ketones. It is also useful in the esterification of carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4′-Dibromoacetophenone is used as a reagent for analysis of fatty acids by HPLC as 4-bromophenacyl esters, for use in the protection of phenols and carboxylic acids. It undergoes condensation reactions with aldehydes in the presence of SnCl2 or SmI3 to afford α,β-unsaturated ketones. It is also useful in the esterification of carboxylic acids.

Solubility
Soluble in dimethyl sulfoxide (5 mg/ml), methanol (20 mg/ml), toluene and ethanol. Insoluble in water.

Notes
Keep container tightly sealed. Store in refrigerated condition. Incompatible with bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text