Thermo Scientific Chemicals

1,3-Dichlorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A13688.30
250 g, Each
Thermo Scientific Chemicals

1,3-Dichlorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A13688.30
250 g, Each
Quantity
Catalog number: A13688.30
also known as A13688-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
541-73-1
IUPAC Name
1,3-dichlorobenzene
Molecular Formula
C6H4Cl2
InChI Key
ZPQOPVIELGIULI-UHFFFAOYSA-N
SMILES
ClC1=CC(Cl)=CC=C1
Assay (GC)
≥97.5%
Refractive Index
1.5435-1.5475 @ 20°C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Identification (FTIR)
Conforms

Description

1,3-Dichlorobenzene is used in medicine, and dyes. And it is also used to study the effect of solvent vapor pressure on the vertical nanowire of C60 molecules1. 1,3-Dichlorobenzene, a chlorinated aromatic hydrocarbon, can be detected in environmental samples using an advanced Fourier transform infrared spectroscopy-attenuated total reflectance (FTIR-ATR) sensor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dichlorobenzene is used in medicine, and dyes. And it is also used to study the effect of solvent vapor pressure on the vertical nanowire of C60 molecules1. 1,3-Dichlorobenzene, a chlorinated aromatic hydrocarbon, can be detected in environmental samples using an advanced Fourier transform infrared spectroscopy-attenuated total reflectance (FTIR-ATR) sensor.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text