Thermo Scientific Chemicals

Acetaldehyde diethyl acetal, 99%, Thermo Scientific Chemicals

Catalog number: A13701.22
100 g, Each
Thermo Scientific Chemicals

Acetaldehyde diethyl acetal, 99%, Thermo Scientific Chemicals

Catalog number: A13701.22
100 g, Each
Quantity
Catalog number: A13701.22
also known as A13701-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
105-57-7
IUPAC Name
1,1-diethoxyethane
Molecular Formula
C6H14O2
InChI Key
DHKHKXVYLBGOIT-UHFFFAOYSA-N
SMILES
CCOC(C)OCC
Identification (FTIR)
Conforms
Form
Liquid
Refractive Index
1.3795-1.3825 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%

Description

Acetaldehyde diethyl acetal is used as a flavoring agent to provide fruit, nut, rum, and whiskey flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetaldehyde diethyl acetal is used as a flavoring agent to provide fruit, nut, rum, and whiskey flavors.

Solubility
Soluble in water (46 g/L at 20°C).

Notes
Keep container tightly sealed. Store in refrigerated condition. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text