Titanium(IV) isopropoxide is used as a precursor for the preparation of titanium and barium-strontium-titanate thin films. It is useful to make porous titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. It is an active component of sharpless epoxidation as well as involved in the synthesis of chiral epoxides. In Kulinkovich reaction, it is involved as a catalyst in the preparation of cyclopropanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Titanium(IV) isopropoxide is used as a precursor for the preparation of titanium and barium-strontium-titanate thin films. It is useful to make porous titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. It is an active component of sharpless epoxidation as well as involved in the synthesis of chiral epoxides. In Kulinkovich reaction, it is involved as a catalyst in the preparation of cyclopropanes.
Solubility
Soluble in anhydrous ethanol, ether, benzene and chloroform.
Notes
Moisture sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids. It reacts with water to produce titanium dioxide.
RUO – Research Use Only
General References:
- Catalyst for transesterification, avoiding acidic or basic conditions. The reaction is carried out such that one alcohol is removed to displace the equilibrium: Synthesis, 138 (1982); Org. Synth. Coll., 8, 201 (1993). For application to carbamates, including Boc to Cbz conversion, see: J. Org. Chem., 62, 7096 (1997).
- For use in the Sharpless enantioselective epoxidation of allylic alcohols, see tert-Butyl hydroperoxide, A13926. For analogous enantioselective oxidation of sulfides to sulfoxides, see Cumene hydroperoxide, L06866. Epoxy alcohols can also be obtained in high yield in one pot by hydroperoxidation of alkenes with singlet oxygen in the presence of Ti(O-i-Pr)4: J. Am. Chem. Soc., 111, 203 (1989).
- Also catalyzes the ring-opening of 2,3-epoxy alcohols (e.g. from the above reactions) with various nucleophiles, including amines, thiols, thiolate anions, halides, carboxylates etc. The mild conditions result in increased regioselectivity: J. Org. Chem., 50, 1557 (1985).
- See also Dimethyl amine hydrochloride, A12133.
- For conversion of epoxides to episulfides, see Thiourea, A12828. For use in reductive alkylation of amines, see Sodium cyanoborohydride, 87839. For use in enantioselective synthesis of homoallylic alcohols, see (R)-(+)-1,1'-Bi(2-naphthol) , L08305. For reduction of amides to aldehydes, see Diphenyl silane, A10884.
- For a brief feature on uses of the reagent, see: Synlett, 2261 (2003).