Thermo Scientific Chemicals

Adipic acid, 99%, Thermo Scientific Chemicals

Catalog number: A13705.0I
5000 g, Each
Thermo Scientific Chemicals

Adipic acid, 99%, Thermo Scientific Chemicals

Catalog number: A13705.0I
5000 g, Each
Quantity
Catalog number: A13705.0I
also known as A13705-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124-04-9
IUPAC Name
hexanedioic acid
Molecular Formula
C6H10O4
InChI Key
WNLRTRBMVRJNCN-UHFFFAOYSA-N
SMILES
OC(=O)CCCCC(O)=O
Identification (FTIR)
Conforms (U.K. specification)
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥98.5% (U.K. specification)
Comment
Purchased in U.K. and U.S.
Melting Point (clear melt)
151-154°C (U.K. specification)

Description

Adipic acid is a monomer used in the production of nylon 66 by polycondenation reaction with hexamethylene diamine. It is also used as a food ingredient and its esters are plasticizers especially in poly vinyl chloride. Further, it reacts with barium hydroxide and undergoes ketonization reaction to get cyclopentanone. In addition to this, it finds application in polyurethane foams, lubricants, adhesives, baking powder, gelling aid and in food flavoring.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adipic acid is a monomer used in the production of nylon 66 by polycondenation reaction with hexamethylene diamine. It is also used as a food ingredient and its esters are plasticizers especially in poly vinyl chloride. Further, it reacts with barium hydroxide and undergoes ketonization reaction to get cyclopentanone. In addition to this, it finds application in polyurethane foams, lubricants, adhesives, baking powder, gelling aid and in food flavoring.

Solubility
Soluble in methanol and acetone. Slightly soluble in cyclohexane. Insoluble in benzene, acetic acid and petroleum ether.

Notes
Incompatible with oxidizing agents
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text