Thermo Scientific Chemicals

Crotonyl chloride, tech. 90%, Thermo Scientific Chemicals

Catalog number: A13710.36
500 g, Each
Thermo Scientific Chemicals

Crotonyl chloride, tech. 90%, Thermo Scientific Chemicals

Catalog number: A13710.36
500 g, Each
Quantity
Catalog number: A13710.36
also known as A13710-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
625-35-4
IUPAC Name
(2E)-but-2-enoyl chloride
Molecular Formula
C4H5ClO
InChI Key
RJUIDDKTATZJFE-NSCUHMNNSA-N
SMILES
C\C=C\C(Cl)=O
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥88.0%
Comment
Remainder mainly cis-isomer
Identification (FTIR)
Conforms
Refractive Index
1.4560-1.4640 @ 20?C

Description

Crotonyl chloride is used in agrochemical, pharmaceutical and dyestuff field. Crotonoyl chloride was used in the synthesis of α,β-unsaturated ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Crotonyl chloride is used in agrochemical, pharmaceutical and dyestuff field. Crotonoyl chloride was used in the synthesis of α,β-unsaturated ester.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store at 4°C. Store away from heat and moisture. Incompatible with
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text