CTS™ Neurobasal™-A Medium
Taking your work to the clinic? Check out CTS B-27 Supplement XenoFree.
CTS™ Neurobasal™-A Medium
Gibco™

CTS™ Neurobasal™-A Medium

CTS Neurobasal-A Medium is designed with the translational stem cell researcher in mind. It can be used to culture post-natalRead more
Have Questions?
Catalog number A1371001
Price (USD)/ 500 mL
143.00
-
Add to cart
Price (USD)/ 500 mL
143.00
Add to cart
CTS™ Neurobasal™-A Medium
Catalog numberA1371001
Price (USD)/ 500 mL
143.00
-
Add to cart

CTS Neurobasal-A Medium is designed with the translational stem cell researcher in mind. It can be used to culture post-natal neurons without the need for an astrocyte feeder layer when used with B-27 supplements.

CTS quality
Gibco CTS products are manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified.

Accompanied by documentation to support your regulatory filing
Specific intended-use statements, full documentation traceability, and convenient access to our Drug Master File (DMF) are available.

For other applications, we provide alternative formulations of CTS Neurobasal media and B-27 supplements.

For established protocols using Neurobasal media, access our Gibco Neurobiology Protocol Handbook.

View publications using Gibco neurobiology products at our Gibco CellCite.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Cell TypeNeural Cells
ClassificationAnimal Origin-free
FormLiquid
Product TypeCell Culture Medium
Serum LevelSerum-free
Shelf Life12 months from date of manufacture
SterilitySterile-filtered
FormatBottle
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing QualityISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity500 mL
Shipping ConditionAmbient
Unit Size500 mL
Contents & Storage
Store at 2–8°C and protect from light.