Thermo Scientific Chemicals

EPPS, 99%, Thermo Scientific Chemicals

Catalog number: A13714.22
100 g, Each
Thermo Scientific Chemicals

EPPS, 99%, Thermo Scientific Chemicals

Catalog number: A13714.22
100 g, Each
Quantity
Catalog number: A13714.22
also known as A13714-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16052-06-5
IUPAC Name
3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
Molecular Formula
C9H20N2O4S
InChI Key
OWXMKDGYPWMGEB-UHFFFAOYSA-N
SMILES
OCCN1CCN(CCCS(O)(=O)=O)CC1
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms

Description

EPPS is used as a biological buffer with a useful pH range of 7.3-8.7 & pKa=8.0 (25°C). It can be used in agrochemical, pharmaceutical and dyestuff field etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
EPPS is used as a biological buffer with a useful pH range of 7.3-8.7 & pKa=8.0 (25°C). It can be used in agrochemical, pharmaceutical and dyestuff field etc.

Solubility
Soluble in water. 25.2 g/L (20°C)

Notes
Store at room temperature. Incompatible with oxidizing agents. Store away from extreme heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text