Kinetin acts as plant growth accelerator, auxin, plant growth regulator, plant cell division promotor. It also acts as cell division factor found in various plant parts and in yeast. A plant growth regulator. Augments growth of microbial cultures.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Kinetin acts as plant growth accelerator, auxin, plant growth regulator, plant cell division promotor. It also acts as cell division factor found in various plant parts and in yeast. A plant growth regulator. Augments growth of microbial cultures.
Solubility
Soluble in water (<1mg/mL) at 25°C, Acetic acid (49.00-51.00 mg/ml), DMSO (43 mg/ml) at 25°C, ethanol (<1mg/mL) at 25°C, 0.1 N Sodium hydroxide (1% w/v), methanol (slightly), and dilute aqueous hydrochloric acid or sodium hydroxide (freely).
Notes
Protect from heat. Store at -20°C. Store away from oxidizing agents.
RUO – Research Use Only
Carlos O. Miller; Folke Skoog; Malcolm H. Von Saltza; F. M. Strong. Kinetin, a Cell Division Factor from Deoxyribonucleic Acid.J. Am. Chem. Soc.1955, 77 (5), 1392-1392.
DJ Osborne. Effect of Kinetin on Protein & Nucleic Acid Metabolism in Xanthium Leaves During Senescence.Plant Physiol.1962, 37 (5), 595-602..
Enhances the rate of cell-division in plants: Plant Physiol., 54, 644 (1974).