Kinetin, 99%, Thermo Scientific Chemicals
Kinetin, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Kinetin, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number A13720.14
Price (USD)/ Each
552.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
552.00
Add to cart
Kinetin, 99%, Thermo Scientific Chemicals
Catalog numberA13720.14
Price (USD)/ Each
552.00
-
Add to cart
Chemical Identifiers
CAS525-79-1
IUPAC NameN-[(furan-2-yl)methyl]-7H-purin-6-amine
Molecular FormulaC10H9N5O
InChI KeyQANMHLXAZMSUEX-UHFFFAOYSA-N
SMILESC(NC1=C2NC=NC2=NC=N1)C1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Identification (FTIR)Conforms
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
FormCrystals or powder or crystalline powder
Assay (HPLC)≥98.5%
Kinetin acts as plant growth accelerator, auxin, plant growth regulator, plant cell division promotor. It also acts as cell division factor found in various plant parts and in yeast. A plant growth regulator. Augments growth of microbial cultures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Kinetin acts as plant growth accelerator, auxin, plant growth regulator, plant cell division promotor. It also acts as cell division factor found in various plant parts and in yeast. A plant growth regulator. Augments growth of microbial cultures.

Solubility
Soluble in water (<1mg/mL) at 25°C, Acetic acid (49.00-51.00 mg/ml), DMSO (43 mg/ml) at 25°C, ethanol (<1mg/mL) at 25°C, 0.1 N Sodium hydroxide (1% w/v), methanol (slightly), and dilute aqueous hydrochloric acid or sodium hydroxide (freely).

Notes
Protect from heat. Store at -20°C. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Carlos O. Miller; Folke Skoog; Malcolm H. Von Saltza; F. M. Strong. Kinetin, a Cell Division Factor from Deoxyribonucleic Acid.J. Am. Chem. Soc.1955, 77 (5), 1392-1392.
  2. DJ Osborne. Effect of Kinetin on Protein & Nucleic Acid Metabolism in Xanthium Leaves During Senescence.Plant Physiol.1962, 37 (5), 595-602..
  3. Enhances the rate of cell-division in plants: Plant Physiol., 54, 644 (1974).