Thermo Scientific Chemicals

2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, 99%, Thermo Scientific Chemicals

Catalog number: A13724.30
250 g, Each
Thermo Scientific Chemicals

2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, 99%, Thermo Scientific Chemicals

Catalog number: A13724.30
250 g, Each
Quantity
Catalog number: A13724.30
also known as A13724-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16357-59-8
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to cream
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Melting Point (clear melt)
61.0-68.0?C

Description

2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline is an irreversible membrane-bound receptor antagonist. It is used in synthetic chemistry, discovery, process R&D. It also acts as coupling reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline is an irreversible membrane-bound receptor antagonist. It is used in synthetic chemistry, discovery, process R&D. It also acts as coupling reagents.

Solubility
Soluble in 95% Ethanol (50 mg/ml). Insoluble in water.

Notes
Moisture Sensitive. Store at 4°C. Incompatible with acids and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text