Thermo Scientific Chemicals

tert-Butyl bromide, 98+%, stab. with potassium carbonate, Thermo Scientific Chemicals

Catalog number: A13727.36
500 g, Each
Thermo Scientific Chemicals

tert-Butyl bromide, 98+%, stab. with potassium carbonate, Thermo Scientific Chemicals

Catalog number: A13727.36
500 g, Each
Quantity
Catalog number: A13727.36
also known as A13727-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
507-19-7
IUPAC Name
2-bromo-2-methylpropane
Molecular Formula
C4H9Br
InChI Key
RKSOPLXZQNSWAS-UHFFFAOYSA-N
SMILES
CC(C)(C)Br
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to very pale yellow
Assay (GC)
≥98.0%
Refractive Index
1.4260-1.4300 @20?C
Form
Liquid

Description

tert-Butyl bromide is used as a raw material in synthetic organic chemistry. It is used to study the massive deadenylation of adenine based-nucleosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl bromide is used as a raw material in synthetic organic chemistry. It is used to study the massive deadenylation of adenine based-nucleosides.

Solubility
Immiscible with water.

Notes
Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text