Thermo Scientific Chemicals

Potassium thiocyanate, 98%, Thermo Scientific Chemicals

Catalog number: A13731.36
500 g, Each
Thermo Scientific Chemicals

Potassium thiocyanate, 98%, Thermo Scientific Chemicals

Catalog number: A13731.36
500 g, Each
Quantity
Catalog number: A13731.36
also known as A13731-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
333-20-0
IUPAC Name
potassium cyanosulfanide
Molecular Formula
CKNS
InChI Key
ZNNZYHKDIALBAK-UHFFFAOYSA-M
SMILES
[K+].[S-]C#N
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
≤2.5%
Appearance (Color)
White
Assay (Argentometric Titration)
≥97.5 to ≤102.5%

Description

Potassium thiocyanate is used in the preparation of ethylenesulfide, lead thiocyante, which is used in the conversion of acyl chloride to thiocyanates. It is also used in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury-thiocyanate and selenocyanate compounds as potential models. It finds application in cosmetic and personal care product formulations as a skin conditioning agent. It is also used in moisturizers and oral care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium thiocyanate is used in the preparation of ethylenesulfide, lead thiocyante, which is used in the conversion of acyl chloride to thiocyanates. It is also used in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury-thiocyanate and selenocyanate compounds as potential models. It finds application in cosmetic and personal care product formulations as a skin conditioning agent. It is also used in moisturizers and oral care products.

Solubility
Soluble in water, acetone and alcohol.

Notes
Hygroscopic. Air, light and moisture sensitive. Incompatible with nitric acids, active halogen compounds and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text