Thermo Scientific Chemicals

2-Aminophenol, 99%, Thermo Scientific Chemicals

Catalog number: A13735.22
100 g, Each
Thermo Scientific Chemicals

2-Aminophenol, 99%, Thermo Scientific Chemicals

Catalog number: A13735.22
100 g, Each
Quantity
Catalog number: A13735.22
also known as A13735-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95-55-6
IUPAC Name
2-aminophenol
Molecular Formula
C6H7NO
InChI Key
CDAWCLOXVUBKRW-UHFFFAOYSA-N
SMILES
NC1=CC=CC=C1O
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
172-177?C
Appearance (Color)
White to grey to brown
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms

Description

2-Aminophenol is used to make dyes and pharmaceuticals. It is also used as a reagent for the synthesis of heterocyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminophenol is used to make dyes and pharmaceuticals. It is also used as a reagent for the synthesis of heterocyclic compounds.

Solubility
Soluble in water (17 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text