Thermo Scientific Chemicals

Geraniol, 97%, Thermo Scientific Chemicals

Catalog number: A13736.18
50 g, Each
Thermo Scientific Chemicals

Geraniol, 97%, Thermo Scientific Chemicals

Catalog number: A13736.18
50 g, Each
Quantity
Catalog number: A13736.18
also known as A13736-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-24-1
IUPAC Name
(2E)-3,7-dimethylocta-2,6-dien-1-ol
Molecular Formula
C10H18O
InChI Key
GLZPCOQZEFWAFX-JXMROGBWSA-N
SMILES
CC(C)=CCCC(C)=CCO
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4745-1.4795 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms

Description

Geraniol is used in the synthesis of insect repellant. It is also used in the synthesis of Angelicoin A and Herecinone J, which inhibit collagen-induced platelet aggregation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Geraniol is used in the synthesis of insect repellant. It is also used in the synthesis of Angelicoin A and Herecinone J, which inhibit collagen-induced platelet aggregation.

Solubility
Not miscible in water.

Notes
Incompatible with Strong oxidizing agents, acid chlorides, acid anhydrides. Keep away from heat. Air sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text