Thermo Scientific Chemicals

Acetone-1,3-dicarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: A13742.18
50 g, Each
Thermo Scientific Chemicals

Acetone-1,3-dicarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: A13742.18
50 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A13742.18
also known as A13742-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
542-05-2
IUPAC Name
3-oxopentanedioate
Molecular Formula
C5H4O5
InChI Key
OXTNCQMOKLOUAM-UHFFFAOYSA-L
SMILES
[O-]C(=O)CC(=O)CC([O-])=O
Loss on Drying
≤2.0% (50?C/constant weight)
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale cream
Assay (Aqueous acid-base Titration)
≥96.0% to ≤104.0%

Description

Acetone-1,3-dicarboxylic acid is used as a building block in organic chemistry. It is also used in the synthesis of hetrocyclic rings and in the Weiss-Cook reaction, which involves the preparation of cis-bicyclo[3.3.0]octane-3,7-dione by reacting with diacyl (1,2 ketone). Its presence in human urine is used as a diagnostic test for the overgrowth of harmful gut flora such as Candida albicans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetone-1,3-dicarboxylic acid is used as a building block in organic chemistry. It is also used in the synthesis of hetrocyclic rings and in the Weiss-Cook reaction, which involves the preparation of cis-bicyclo[3.3.0]octane-3,7-dione by reacting with diacyl (1,2 ketone). Its presence in human urine is used as a diagnostic test for the overgrowth of harmful gut flora such as Candida albicans.

Solubility
Soluble in water and alcohol. Slightly soluble in ethyl acetate and ether. Insoluble in chloroform.

Notes
Hygroscopic. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text