Gibco™ YPD Broth is a ready-to-use liquid growth media used for yeast culture. Our filter-sterilized Gibco™ YPD Broth can beRead more
Have Questions?
Catalog Number
Quantity
A1374501
1 L
Catalog number A1374501
Price (USD)
57.65
Online Exclusive
60.00
Save 2.35 (4%)
Each
In stock
Add to cart
Quantity:
1 L
Price (USD)
57.65
Online Exclusive
60.00
Save 2.35 (4%)
Each
Add to cart
Gibco™ YPD Broth is a ready-to-use liquid growth media used for yeast culture. Our filter-sterilized Gibco™ YPD Broth can be used for nearly all research applications that require a liquid yeast culturing step such as protein expression, cloning, and expansion. • Gibco™ microbiological media • Convenient ready-to-use liquid format • 1 L size available
YPD Broth Recipe YPD Broth is a commonly used yeast growth media for maintenance and propagation of Pichia pastoris (P. pastoris) and Saccharomyces cerevisae (S. cerevisae) yeast strains.
Formulation per One Liter: 10 g Yeast extract 20 g Bacto peptone 20 g Dextrose (glucose)
Ready-to-Use in Optimized Gibco™ Media Bottles Each bottle of Gibco™ YPD Broth is filter-sterilized and ready-to-use in ergonomic Gibco™ media bottles with angled openings for simplified pipetting.
For research use only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeLiquid Medium
Media TypeYPD
Preparation MethodReady-to-Use
Product LineGibco™
Quantity1 L
Shipping ConditionRoom Temperature
FormLiquid
Product TypeMicrobiology Media
SterilitySterile
Target Organism ClassP. pastoris, S. cerevisiae
Unit SizeEach
Contents & Storage
1 liter YPD Broth, liquid; store at 2 to 8 °C. Protect from light.