Thermo Scientific Chemicals

2,4-Dihydroxybenzophenone, 99%, Thermo Scientific Chemicals

Catalog number: A13749.0E
2500 g, Each
Thermo Scientific Chemicals

2,4-Dihydroxybenzophenone, 99%, Thermo Scientific Chemicals

Catalog number: A13749.0E
2500 g, Each
Quantity
Catalog number: A13749.0E
also known as A13749-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
131-56-6
IUPAC Name
4-benzoylbenzene-1,3-diol
Molecular Formula
C13H10O3
InChI Key
ZXDDPOHVAMWLBH-UHFFFAOYSA-N
SMILES
OC1=CC=C(C(=O)C2=CC=CC=C2)C(O)=C1
Appearance (Color)
White to pale cream to pale yellow
Assay (GC)
≥98.5%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
141.0-149.0?C
Identification (FTIR)
Conforms

Description

Novel mordent and disperse azo dyes were prepared by the coupling of various diazo solutions of aromatic amines with 2, 4-dihydroxybenzophenone. The ultraviolet absorbing monomers were synthesized by reaction of 2, 4-dihydroxybenzophenone with glycidyl acrylate and glycidyl methacrylate. Two new polymerizable stabilizers, 4-benzoyl-2-(α-piperidino-2-chlorobenzyl)-3-hydroxyphenyl acrylate (BPBHA) and corresponding methacrylate (BPBHMA), were synthesized and characterized from 2,4-Dihydroxybenzophenone. a polymerizable UV-stabilizer, 2-hydroxy-4-(3-methacryloxy-2-hydroxylpropoxy) benzophenone (BPMA), was synthesized using 2, 4-dihydroxybenzophenone (UV-0) and glycidyl methacrylate (GMA).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Novel mordent and disperse azo dyes were prepared by the coupling of various diazo solutions of aromatic amines with 2, 4-dihydroxybenzophenone. The ultraviolet absorbing monomers were synthesized by reaction of 2, 4-dihydroxybenzophenone with glycidyl acrylate and glycidyl methacrylate. Two new polymerizable stabilizers, 4-benzoyl-2-(α-piperidino-2-chlorobenzyl)-3-hydroxyphenyl acrylate (BPBHA) and corresponding methacrylate (BPBHMA), were synthesized and characterized from 2,4-Dihydroxybenzophenone. a polymerizable UV-stabilizer, 2-hydroxy-4-(3-methacryloxy-2-hydroxylpropoxy) benzophenone (BPMA), was synthesized using 2, 4-dihydroxybenzophenone (UV-0) and glycidyl methacrylate (GMA).

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text