3,5-Dibromotoluene, 98+%, Thermo Scientific Chemicals
3,5-Dibromotoluene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dibromotoluene, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A13756.22
also known as A13756-22
Price (USD)/ Each
629.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
629.00
Add to cart
3,5-Dibromotoluene, 98+%, Thermo Scientific Chemicals
Catalog numberA13756.22
Price (USD)/ Each
629.00
-
Add to cart
Chemical Identifiers
CAS1611-92-3
IUPAC Name1,3-dibromo-5-methylbenzene
Molecular FormulaC7H6Br2
InChI KeyDPKKOVGCHDUSAI-UHFFFAOYSA-N
SMILESCC1=CC(Br)=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Appearance (Color)White to pale cream to cream to yellow
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)37.0-43.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only