4-Aminobenzophenone is associated with 4-benzoylpyridine and it is used as a frequency doubler for semiconductor lasers. It is used in the preparation of 1-[4-[N,N-bis(trimethylsilyl)amino]phenyl]-1-phenylethylene. It is used as a precursor to prepare 1,2,3-triazoles, pyrimidotriazoles, benzothiazole and pyrimidinone derivatives. It is involved in the formation of coordination complexes with chromium(III).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Aminobenzophenone is associated with 4-benzoylpyridine and it is used as a frequency doubler for semiconductor lasers. It is used in the preparation of 1-[4-[N,N-bis(trimethylsilyl)amino]phenyl]-1-phenylethylene. It is used as a precursor to prepare 1,2,3-triazoles, pyrimidotriazoles, benzothiazole and pyrimidinone derivatives. It is involved in the formation of coordination complexes with chromium(III).
Solubility
Soluble in cold water, alcohol and ether.
Notes
Air and light sensitive. Incompatible with strong acids.
RUO – Research Use Only
Natarajan, V.; Usharani, S.; Arivanandhan, M.; Anandan, P.; Hayakawa, Y. Effect of solvents on the bulk growth of 4-aminobenzophenone single crystals: A potential material for blue and green lasers. Spectrochim. Acta, Part A 2015, 145, 329-332.
Rajendiran, N.; Sankaranarayanan, R. K. Nanoscale self assembly of cyclodextrin capped 4-aminobenzophenone via non-covalent interactions. J. Mol. Liq. 2015, 206, 218-227.