Thermo Scientific Chemicals

4-Aminobenzophenone, 98%, Thermo Scientific Chemicals

Catalog number: A13757.18
50 g, Each
Thermo Scientific Chemicals

4-Aminobenzophenone, 98%, Thermo Scientific Chemicals

Catalog number: A13757.18
50 g, Each
Quantity
Catalog number: A13757.18
also known as A13757-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1137-41-3
Appearance (Color)
Cream to yellow to brown or purple
Assay (GC)
≥97.5%
Form
Powder
Identification (FTIR)
Conforms

Description

4-Aminobenzophenone is associated with 4-benzoylpyridine and it is used as a frequency doubler for semiconductor lasers. It is used in the preparation of 1-[4-[N,N-bis(trimethylsilyl)amino]phenyl]-1-phenylethylene. It is used as a precursor to prepare 1,2,3-triazoles, pyrimidotriazoles, benzothiazole and pyrimidinone derivatives. It is involved in the formation of coordination complexes with chromium(III).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzophenone is associated with 4-benzoylpyridine and it is used as a frequency doubler for semiconductor lasers. It is used in the preparation of 1-[4-[N,N-bis(trimethylsilyl)amino]phenyl]-1-phenylethylene. It is used as a precursor to prepare 1,2,3-triazoles, pyrimidotriazoles, benzothiazole and pyrimidinone derivatives. It is involved in the formation of coordination complexes with chromium(III).

Solubility
Soluble in cold water, alcohol and ether.

Notes
Air and light sensitive. Incompatible with strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text