Thermo Scientific Chemicals

Trimethyl orthoformate, 99%, Thermo Scientific Chemicals

Catalog number: A13760.AP
500 mL, Each
Thermo Scientific Chemicals

Trimethyl orthoformate, 99%, Thermo Scientific Chemicals

Catalog number: A13760.AP
500 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A13760.AP
also known as A13760-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
149-73-5
IUPAC Name
trimethoxymethane
Molecular Formula
C4H10O3
InChI Key
PYOKUURKVVELLB-UHFFFAOYSA-N
SMILES
COC(OC)OC
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless
Form
Liquid
Refractive Index
1.3775-1.3805 @ 20?C
Identification (FTIR)
Conforms

Description

Trimethyl orthoformate is used as a protecting group for aldehydes in organic synthesis, as an additive in polyurethane coatings and as a dehydrating agent in the preparation of surface modified colloidal silica nanoparticles. It is also used as a chemical intermediate in the preparation of vitamin B1 and sulfa drugs. It acts as an effective solvent for thallium(III) nitrate-mediated oxidations. Furthermore. It is utilized for the synthesis of chromone from keto-hydroxy naphthol in the presence of trimethylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethyl orthoformate is used as a protecting group for aldehydes in organic synthesis, as an additive in polyurethane coatings and as a dehydrating agent in the preparation of surface modified colloidal silica nanoparticles. It is also used as a chemical intermediate in the preparation of vitamin B1 and sulfa drugs. It acts as an effective solvent for thallium(III) nitrate-mediated oxidations. Furthermore. It is utilized for the synthesis of chromone from keto-hydroxy naphthol in the presence of trimethylamine.

Solubility
Miscible with ether, alcohol and benzene.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text