Thermo Scientific Chemicals

L-Cystine, 99%, Thermo Scientific Chemicals

Catalog number: A13762.18
50 g, Each
Thermo Scientific Chemicals

L-Cystine, 99%, Thermo Scientific Chemicals

Catalog number: A13762.18
50 g, Each
Quantity
Catalog number: A13762.18
also known as A13762-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-89-3
IUPAC Name
(2R)-2-amino-3-{[(2R)-2-amino-2-carboxyethyl]disulfanyl}propanoic acid
Molecular Formula
C6H12N2O4S2
InChI Key
LEVWYRKDKASIDU-IMJSIDKUSA-N
SMILES
N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Optical Rotation
-210° to -224° (C=1 in 1N HCl)

Description

L-Cystine is used as an antioxidant, protecting tissues against radiation and pollution. It finds application in protein synthesis. It is required for utilization of vitamin B6 and is useful in healing burns and wounds. It is also is required by certain malignant cell lines in the culture medium as well as for the growth of certain micro-organisms. It is useful in the stimulation of hematopoietic system and promotes the formation of white and red blood cells. It is an active ingredient in medications used to treat dermatitis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text