Thermo Scientific Chemicals

Methyl trimethylacetate, 99%, Thermo Scientific Chemicals

Catalog number: A13764.36
500 g, Each
Thermo Scientific Chemicals

Methyl trimethylacetate, 99%, Thermo Scientific Chemicals

Catalog number: A13764.36
500 g, Each
Quantity
Catalog number: A13764.36
also known as A13764-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
598-98-1
IUPAC Name
methyl 2,2-dimethylpropanoate
Molecular Formula
C6H12O2
InChI Key
CNMFHDIDIMZHKY-UHFFFAOYSA-N
SMILES
COC(=O)C(C)(C)C
Refractive Index
1.3885-1.3915 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text