Thermo Scientific Chemicals

4-Methylbenzylamine, 98%, Thermo Scientific Chemicals

Catalog number: A13768.14
25 g, Each
Thermo Scientific Chemicals

4-Methylbenzylamine, 98%, Thermo Scientific Chemicals

Catalog number: A13768.14
25 g, Each
Quantity
Catalog number: A13768.14
also known as A13768-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
104-84-7
IUPAC Name
1-(4-methylphenyl)methanamine
Molecular Formula
C8H11N
InChI Key
HMTSWYPNXFHGEP-UHFFFAOYSA-N
SMILES
CC1=CC=C(CN)C=C1
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.5330-1.5370 @ 20?C
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Form
Liquid

Description

The reactions of 1-hexanol with benzylamine and with 4-methylbenzylamine led to the corresponding imines in moderate yields. 4-methylbenzylamine in ethanol/water solution (1:4) as simultaneously absorbing and buffering background electrolyte with detection at 210 nm was found suitable for determination of the individual compounds in determination by capillary zone electrophoresis with indirect detection. Synthesis of benzimidazoles uses 4-methylbenzylamine (R 1 =4-MeBn) and 4-chlorophenyl isothiocyanate (R 2 =4-ClPh) as building blocks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The reactions of 1-hexanol with benzylamine and with 4-methylbenzylamine led to the corresponding imines in moderate yields. 4-methylbenzylamine in ethanol/water solution (1:4) as simultaneously absorbing and buffering background electrolyte with detection at 210 nm was found suitable for determination of the individual compounds in determination by capillary zone electrophoresis with indirect detection. Synthesis of benzimidazoles uses 4-methylbenzylamine (R 1 =4-MeBn) and 4-chlorophenyl isothiocyanate (R 2 =4-ClPh) as building blocks.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text