Thermo Scientific Chemicals

4-Allyloxybenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A13773.18
50 g, Each
Thermo Scientific Chemicals

4-Allyloxybenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A13773.18
50 g, Each
Quantity
Catalog number: A13773.18
also known as A13773-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
40663-68-1
IUPAC Name
4-(prop-2-en-1-yloxy)benzaldehyde
Molecular Formula
C10H10O2
InChI Key
TYNJQOJWNMZQFZ-UHFFFAOYSA-N
SMILES
C=CCOC1=CC=C(C=O)C=C1
Assay (GC)
≥96.0%
Refractive Index
1.5660-1.5710 @ 20?C
Form
Liquid
Free acid (titration)
≤1.5%
Appearance (Color)
Clear colorless to pale yellow

Description

4-Allyloxybenzaldehyde is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Allyloxybenzaldehyde is used as pharmaceutical intermediates.

Solubility
Immiscible in water.

Notes
Use with adequate ventilation. Store under inert atmosphere. Protect from moisture and heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text