Thermo Scientific Chemicals

1-Chloro-2,4-dinitrobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A13774.36
500 g, Each
Thermo Scientific Chemicals

1-Chloro-2,4-dinitrobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A13774.36
500 g, Each
Quantity
Catalog number: A13774.36
also known as A13774-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
97-00-7
Appearance (Color)
Pale yellow to yellow to orange to black
Assay (GC)
≥97.5%
Melting Point (clear melt)
46.0-53.0?C
Form
Crystals or powder or crystalline powder or lumps or fused solid or flakes
Identification (FTIR)
Conforms

Description

1-Chloro-2,4-dinitrobenzene is used as a reagent for the detection and determination of pyridine compounds. It has been used as alkylating agent to evaluate the depletion of intracellular erythrocyte glutathione (GSH). It is an irreversible inhibitor of human thioredoxin reductase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2,4-dinitrobenzene is used as a reagent for the detection and determination of pyridine compounds. It has been used as alkylating agent to evaluate the depletion of intracellular erythrocyte glutathione (GSH). It is an irreversible inhibitor of human thioredoxin reductase.

Solubility
Soluble in chloroform, ether, hot alcohol, and benzene. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, ammonia. Reacts with hydrazine hydrate.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text