Thermo Scientific™

Diethyl ethoxymethylenemalonate, 98+%, Thermo Scientific Chemicals

Catalog number: A13776.18
50 g, Each
Thermo Scientific™

Diethyl ethoxymethylenemalonate, 98+%, Thermo Scientific Chemicals

Catalog number: A13776.18
50 g, Each
Quantity
Catalog number: A13776.18
also known as A13776-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
87-13-8
IUPAC Name
1,3-diethyl 2-(ethoxymethylidene)propanedioate
Molecular Formula
C10H16O5
InChI Key
LTMHNWPUDSTBKD-UHFFFAOYSA-N
SMILES
CCOC=C(C(=O)OCC)C(=O)OCC
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Assay (GC)
≥98.0%
Refractive Index
1.4605-1.4645 @ 20?C
Form
Liquid

Description

Diethyl ethoxymethylenemalonate is used for synthesis of pyrido[3,2-e]pyrimido[1,2-c]pyrimidines. Initially, it was used to monitor lysine decarboxylase activity. It is useful in the determination of amino acids by precolumn derivatization by using reversed-phase high-performance liquid chromatography (HPLC). It plays an important role in the Gould-Jacobs reaction to prepare quinolines. For example, anile reacts with this reagent to give 4-hydroxyquinoline. It acts as an intermediate in the production of flumequine, which is a fluoroquinolone antibiotic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl ethoxymethylenemalonate is used for synthesis of pyrido[3,2-e]pyrimido[1,2-c]pyrimidines. Initially, it was used to monitor lysine decarboxylase activity. It is useful in the determination of amino acids by precolumn derivatization by using reversed-phase high-performance liquid chromatography (HPLC). It plays an important role in the Gould-Jacobs reaction to prepare quinolines. For example, anile reacts with this reagent to give 4-hydroxyquinoline. It acts as an intermediate in the production of flumequine, which is a fluoroquinolone antibiotic.

Solubility
Miscible with chloroform, dichloromethane and methanol. Immiscible with water.

Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text