Thermo Scientific Chemicals

Phenylphosphonic dichloride, 90+%, Thermo Scientific Chemicals

Catalog number: A13777.30
250 g, Each
Thermo Scientific Chemicals

Phenylphosphonic dichloride, 90+%, Thermo Scientific Chemicals

Catalog number: A13777.30
250 g, Each
Quantity
Catalog number: A13777.30
also known as A13777-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
824-72-6
IUPAC Name
phenylphosphonoyl dichloride
Molecular Formula
C6H5Cl2OP
InChI Key
IBDMRHDXAQZJAP-UHFFFAOYSA-N
SMILES
ClP(Cl)(=O)C1=CC=CC=C1
Refractive Index
1.5190-1.5990 @ 20?C
Assay (GC)
≥90.0%
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow

Description

Phenylphosphonic dichloride is used in the preparation of 2-phenyl-[1,3,2]dioxaphosphepane-2-oxide. Further, it acts as an effective reagent for the chlorinative dehydration of some heterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylphosphonic dichloride is used in the preparation of 2-phenyl-[1,3,2]dioxaphosphepane-2-oxide. Further, it acts as an effective reagent for the chlorinative dehydration of some heterocycles.

Solubility
Miscible with benzene, chloroform, dimethyl sulfoxide and carbon tetrachloride.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong bases and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text