Glycylglycylglycine, 98+%
Glycylglycylglycine, 98+%
Thermo Scientific Chemicals

Glycylglycylglycine, 98+%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number A13778.06
also known as A13778-06
Price (USD)/ Each
68.65
Online exclusive
75.90 
Save 7.25 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
68.65
Online exclusive
75.90 
Save 7.25 (10%)
Add to cart
Glycylglycylglycine, 98+%
Catalog numberA13778.06
Price (USD)/ Each
68.65
Online exclusive
75.90 
Save 7.25 (10%)
-
Add to cart
Chemical Identifiers
CAS556-33-2
IUPAC Name2-[2-(2-aminoacetamido)acetamido]acetic acid
Molecular FormulaC6H11N3O4
InChI KeyXKUKSGPZAADMRA-UHFFFAOYSA-N
SMILESNCC(=O)NCC(=O)NCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)White
FormPowder
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0% (non-U.S. specification)
Assay from Supplier's CofA≥98.0% (U.S. specification)
View more
Glycylglycylglycine is used as a model peptide for studies of physicochemical parameters and molecular associations of small peptides. Triglycine is used as a copper chelator. It also acts as substrate for reproducible serum protein measurements by the biuret reaction1 and for the assay of aminotripeptidases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycylglycylglycine is used as a model peptide for studies of physicochemical parameters and molecular associations of small peptides. Triglycine is used as a copper chelator. It also acts as substrate for reproducible serum protein measurements by the biuret reaction1 and for the assay of aminotripeptidases

Solubility
Soluble in water (94.6 mg/ml at 20°C), methanol (50 mg/ml), and 80% acetic acid (50 mg/ml).

Notes
Store at -20°C. Protect from moisture. Avoid excessive heat and light.
RUO – Research Use Only

General References:

  1. MK Kumaran; ID Watson and GR Hedwig. Thermodynamic properties of peptide solutions. The relative partial molar enthalpies of aqueous solutions of glycylglycylglycine, glycylglycylalanine and alanylglycylglycine.Australian Journal of Chemistry.1983, 36 (9),1813 - 1820 .
  2. H. L. Yakel and E. W. Hughes. The unit-cell dimensions and space groups of two modifications of crystalline glycylglycylglycine.Acta Cryst.1952, 5 847-848 .