1-Iodohexane, 98%, stab. with copper, Thermo Scientific Chemicals
1-Iodohexane, 98%, stab. with copper, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Iodohexane, 98%, stab. with copper, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A13784.14
also known as A13784-14
Price (USD)/ Each
42.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
42.00
Add to cart
1-Iodohexane, 98%, stab. with copper, Thermo Scientific Chemicals
Catalog numberA13784.14
Price (USD)/ Each
42.00
-
Add to cart
Chemical Identifiers
CAS638-45-9
IUPAC Name1-iodohexane
Molecular FormulaC6H13I
InChI KeyANOOTOPTCJRUPK-UHFFFAOYSA-N
SMILESCCCCCCI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.4900-1.4940 @ 20?C
FormLiquid
CommentStabilised with copper powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only