Thermo Scientific Chemicals

m-Toluic acid, 99%, Thermo Scientific Chemicals

Catalog number: A13785.0B
1000 g, Each
Thermo Scientific Chemicals

m-Toluic acid, 99%, Thermo Scientific Chemicals

Catalog number: A13785.0B
1000 g, Each
Quantity
Catalog number: A13785.0B
also known as A13785-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-04-7
Appearance (Color)
White to pale yellow
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Assay (Silylated GC)
≥98.5%
Melting Point (clear melt)
108.0-115.0°C
Form
Crystals or powder or crystalline powder or flakes

Description

m-Toluic acid is used for the production of N,N-di methyl formamide, toluene formyl chloride and toluene nitrile etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Toluic acid is used for the production of N,N-di methyl formamide, toluene formyl chloride and toluene nitrile etc,.

Solubility
Soluble in alcohol and ether.

Notes
Incompatible with strong oxidizing agents. Store away from incompatible materials such as oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text