m-Toluic acid is used for the production of N,N-di methyl formamide, toluene formyl chloride and toluene nitrile etc,.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
m-Toluic acid is used for the production of N,N-di methyl formamide, toluene formyl chloride and toluene nitrile etc,.
Solubility
Soluble in alcohol and ether.
Notes
Incompatible with strong oxidizing agents. Store away from incompatible materials such as oxidizing agents.
RUO – Research Use Only
Yong Ge; Frederic H Vaillancourt; Nathalie Y R Agar, Lindsay D Eltis. Reactivity of toluate dioxygenase with substituted benzoates and dioxygen. Journal of Bacteriology.2002, 184 (15), 4096-4103
I Sarand, S Timonen, T Koivula, R Peltola, K Haahtela, R Sen, M Romantschuk. Tolerance and biodegradation of m-toluate by Scots pine, a mycorrhizal fungus and fluorescent pseudomonads individually and under associative conditions.Journal of Applied Microbiology.1999, 86 (5), 817-826