Thermo Scientific™

4-Methoxybenzenesulfonyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A13788.22
100 g, Each
Thermo Scientific™

4-Methoxybenzenesulfonyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A13788.22
100 g, Each
Quantity
Catalog number: A13788.22
also known as A13788-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-68-0
IUPAC Name
4-methoxybenzene-1-sulfonyl chloride
Molecular Formula
C7H7ClO3S
InChI Key
DTJVECUKADWGMO-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=C1)S(Cl)(=O)=O
Appearance (Color)
White to pale cream
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%
Melting Point (clear melt)
36.0-43.0?C
Form
Crystals or powder or crystalline powder or lumps or chunks or fused solid
Assay (GC)
≥98.0%

Description

4-Methoxybenzenesulfonyl chloride is used as an anti-HIV as Pharmaceutical Intermediates. It also acts as protecting group agent for various nitrogen functions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzenesulfonyl chloride is used as an anti-HIV as Pharmaceutical Intermediates. It also acts as protecting group agent for various nitrogen functions.

Solubility
Soluble in Toluene. Decomposes in water.

Notes
Moisture Sensitive. Store at 4°C. Store in a cool and dark place. Protect from moisture. Store away from strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text