Thermo Scientific Chemicals

2-Bromopyrimidine, 98+%, Thermo Scientific Chemicals

Catalog number: A13791.22
100 g, Each
Thermo Scientific Chemicals

2-Bromopyrimidine, 98+%, Thermo Scientific Chemicals

Catalog number: A13791.22
100 g, Each
Quantity
Catalog number: A13791.22
also known as A13791-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4595-60-2
IUPAC Name
2-bromopyrimidine
Molecular Formula
C4H3BrN2
InChI Key
PGFIHORVILKHIA-UHFFFAOYSA-N
SMILES
BrC1=NC=CC=N1
Appearance (Color)
White to cream or pale yellow or pale brown
Assay (GC)
≥98.0% (non-U.S. specification)
Assay from Suppliers CofA
≥98.0% (U.S. specification)
Comment
Purchased in the U.S. and in other countries
Identification (FTIR)
Conforms (non-U.S. specification)

Description

2-Bromopyrimidine is used in the preparation of 2,7-bis(2-pyrimidyl)-9,9-dihexylfluorene by cross-coupling with 9,9-dihexylfluorene-2,7-diboronic acid. It undergoes microwave-assisted aminocarbonylation at C-2 with palladium and molybdenum hexacarbonyl.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopyrimidine is used in the preparation of 2,7-bis(2-pyrimidyl)-9,9-dihexylfluorene by cross-coupling with 9,9-dihexylfluorene-2,7-diboronic acid. It undergoes microwave-assisted aminocarbonylation at C-2 with palladium and molybdenum hexacarbonyl.

Solubility
Miscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids, acid chlorides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text