Gibco™

BG-11 Media, optimized for cyanobacteria

Catalog number: A1379901
Have Questions?
Gibco™

BG-11 Media, optimized for cyanobacteria

Catalog number: A1379901
Have Questions?
Catalog Number
A1379901
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
6 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A13799011 L
-
A13799026 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Gibco™ BG-11 Media is optimized for the growth and maintenance of select Cyanobacteria including Synechococcus elongates. The formulation comes ready-to-use at 1X to let you avoid laborious media preparation steps.

Each Gibco™ BG-11 Media comes in the award winning Gibco™ bottle designed for easier use in the biosafety cabinet, minimizing the risk of contamination, and helps you perform cell culture more consistently. Together, the superior packaging and quality, greater reliability, and improved consistency in Cyanobacteria culture, results in better overall efficiency and more robust data (Fig. 1, 2).

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.

Specifications

Form
Liquid
Media Type
Algal Media
Preparation Method
None (Ready-to-Use)
Product Type
Microbiology Media
Target Organism Class
Cyanobacteria
Green Features
Sustainable packaging
Quantity
1 L
Shipping Condition
Room Temperature

Contents & Storage

Gibco™ BG-11 Growth Media contains:

•  1 L Gibco™ BG-11 Media — Store at 4 °C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text