Thermo Scientific Chemicals

2,2-Dimethoxypropane, 98%, Thermo Scientific Chemicals

Catalog number: A13810.AE
100 mL, Each
Thermo Scientific Chemicals

2,2-Dimethoxypropane, 98%, Thermo Scientific Chemicals

Catalog number: A13810.AE
100 mL, Each
Quantity
Catalog number: A13810.AE
also known as A13810-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-76-9
IUPAC Name
2,2-dimethoxypropane
Molecular Formula
C5H12O2
InChI Key
HEWZVZIVELJPQZ-UHFFFAOYSA-N
SMILES
COC(C)(C)OC
Assay (GC)
≥97.5%
Refractive Index
1.3755-1.3795 @ 20?C (UK sourced material only)
Appearance (Color)
Clear colorless
Form
Liquid
Identification (FTIR)
Conforms (UK sourced material only)

Description

2,2-Dimethoxypropane acts as a dehydrating agent. It also serves as an intermediate in the synthesis of vitamin E, vitamin A and various carotenoids such as astaxanthin. It is used as a reagent for the preparation of 1,2-diols, acetonides, isopropylidene derivatives of sugars, nucleosides, methyl esters of amino acids and enol ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dimethoxypropane acts as a dehydrating agent. It also serves as an intermediate in the synthesis of vitamin E, vitamin A and various carotenoids such as astaxanthin. It is used as a reagent for the preparation of 1,2-diols, acetonides, isopropylidene derivatives of sugars, nucleosides, methyl esters of amino acids and enol ethers.

Solubility
Soluble in benzene, carbon tetrachloride, ethyl ether and n-butane, methanol. Moderately soluble in water.

Notes
Incompatible with oxidizing agents and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text