Thermo Scientific Chemicals

10-Undecynoic acid, 96%, Thermo Scientific Chemicals

Catalog number: A13815.06
5 g, Each
Thermo Scientific Chemicals

10-Undecynoic acid, 96%, Thermo Scientific Chemicals

Catalog number: A13815.06
5 g, Each
Quantity
Catalog number: A13815.06
also known as A13815-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2777-65-3
IUPAC Name
undec-10-ynoate
Molecular Formula
C11H17O2
InChI Key
OAOUTNMJEFWJPO-UHFFFAOYSA-M
SMILES
[O-]C(=O)CCCCCCCCC#C
Form
Powder
Assay (Silylated GC)
≥95.0%
Melting Point (clear melt)
40-45?C
Appearance (Color)
White to cream to pale yellow or pale brown
Assay (Aqueous acid-base Titration)
≥95.0% to ≤105.0%

Description

10-Undecynoic acid may be employed as a biochemical probe in an assay for the microsomal hydroxylation of lauric acid (LA), based on HPLC with flow-through radiochemical detection. It was employed as model compound to investigate the microwave assisted surface click reactions catalyzed with Cu(II)/sodium L-ascorbate. It is reported as highly selective irreversible inhibitor of hepatic ω- and ω-1-lauric acid hydroxylases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
10-Undecynoic acid may be employed as a biochemical probe in an assay for the microsomal hydroxylation of lauric acid (LA), based on HPLC with flow-through radiochemical detection. It was employed as model compound to investigate the microwave assisted surface click reactions catalyzed with Cu(II)/sodium L-ascorbate. It is reported as highly selective irreversible inhibitor of hepatic ω- and ω-1-lauric acid hydroxylases.

Solubility
Sparingly Soluble in water.

Notes
Store at 4°C. Store away from incompatible materials such as oxidizing agents
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text