Thermo Scientific Chemicals

Glycine, 99%

Catalog number: A13816.0E
2500 g, Each
Thermo Scientific Chemicals

Glycine, 99%

Catalog number: A13816.0E
2500 g, Each
Quantity
Catalog number: A13816.0E
also known as A13816-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-40-6
IUPAC Name
2-aminoacetic acid
Molecular Formula
C2H5NO2
InChI Key
DHMQDGOQFOQNFH-UHFFFAOYSA-N
SMILES
NCC(O)=O
Appearance (Color)
White to pale cream
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5% (non-U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated
Form
Crystals or powder or crystalline powder
Assay from Suppliers CofA
≥98.5% (U.S. specification)

Description

Glycine is used as an additive in pet food, animal feed and sweetener and taste enhancer in human foods. It is also used as a buffering agent in antacids, analgesics, antiperspirants, cosmetics and toiletries. It acts as a neurotransmitter in the central nervous system, especially in the spinal cord, brainstem, and retina. It serves as a buffer agent and prevents the sample damage during electrophoresis. Further, it is used to remove protein-labelling antibodies from western blot membranes. In addition to this, it is involved in the production of rubber sponge products, metal complexants and fertilizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine is used as an additive in pet food, animal feed and sweetener and taste enhancer in human foods. It is also used as a buffering agent in antacids, analgesics, antiperspirants, cosmetics and toiletries. It acts as a neurotransmitter in the central nervous system, especially in the spinal cord, brainstem, and retina. It serves as a buffer agent and prevents the sample damage during electrophoresis. Further, it is used to remove protein-labelling antibodies from western blot membranes. In addition to this, it is involved in the production of rubber sponge products, metal complexants and fertilizers.

Solubility
Soluble in water and pyridine. Slightly soluble in acetone. Insoluble in diethyl ether, n-octanol and ethanol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text