Thermo Scientific Chemicals

n-Butyl L-lactate, 97+%, Thermo Scientific Chemicals

Catalog number: A13821.30
250 g, Each
Thermo Scientific Chemicals

n-Butyl L-lactate, 97+%, Thermo Scientific Chemicals

Catalog number: A13821.30
250 g, Each
Quantity
Catalog number: A13821.30
also known as A13821-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
34451-19-9
Identification (FTIR)
Conforms
Optical Rotation
-11.0 ± 1.9° (Neat)
Form
Liquid
Appearance (Color)
Clear colorless
Refractive Index
1.4185-1.4235 @ 20°C

Description

Fenofibrate was formulated with n-butyl L-lactate, Tween 80, and a number of cosurfactants, diluted with the water phase (either water or saccharidesolution) and then subjected to a freeze-drying (FD) process to obtain SMEPMS nanosized.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fenofibrate was formulated with n-butyl L-lactate, Tween 80, and a number of cosurfactants, diluted with the water phase (either water or saccharidesolution) and then subjected to a freeze-drying (FD) process to obtain SMEPMS nanosized.

Solubility
Soluble in alcohol and water(42 g/L (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text