Thermo Scientific Chemicals

2-Butyne, 98%, Thermo Scientific Chemicals

Catalog number: A13823.14
25 g, Each
Thermo Scientific Chemicals

2-Butyne, 98%, Thermo Scientific Chemicals

Catalog number: A13823.14
25 g, Each
Quantity
Catalog number: A13823.14
also known as A13823-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
503-17-3
IUPAC Name
but-2-yne
Molecular Formula
C4H6
InChI Key
XNMQEEKYCVKGBD-UHFFFAOYSA-N
SMILES
CC#CC
Appearance (Color)
Clear colorless to very pale yellow
Refractive Index
1.3900-1.3940 @ 20?C
Form
Liquid
Assay (GC)
≥97.5%

Description

2-Butyne is used to synthesize alkylated hydroquinones in the total synthesis of Vitamin E. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Butyne is used to synthesize alkylated hydroquinones in the total synthesis of Vitamin E. It is also used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Avoid extremes of temperature and direct sunlight. Heat-sensitive. Avoid contact with oxidizing materials. Incompatible with Halogens. Chlorinated solvents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text