Thermo Scientific Chemicals

(1-Hexyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals

Catalog number: A13826.22
100 g, Each
Thermo Scientific Chemicals

(1-Hexyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals

Catalog number: A13826.22
100 g, Each
Quantity
Catalog number: A13826.22
also known as A13826-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4762-26-9
IUPAC Name
hexyltriphenylphosphanium bromide
Molecular Formula
C24H28BrP
InChI Key
PWDFZWZPWFYFTC-UHFFFAOYSA-M
SMILES
[Br-].CCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Form
Powder or lumps
Melting Point (clear melt)
198.0-205.0?C
Assay (Titration ex Bromide)
≥97.5 to ≤102.5%
Appearance (Color)
White to pale cream to pale yellow
Water Content (Karl Fischer Titration)
≤1%

Description

(1-Hexyl)triphenylphosphonium bromide can be used as catalyst in pharmaceutical research. It is used in organic synthesis. Reactive wittg reagent used in prostaglandin synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Hexyl)triphenylphosphonium bromide can be used as catalyst in pharmaceutical research. It is used in organic synthesis. Reactive wittg reagent used in prostaglandin synthesis.

Solubility
Soluble in water and methanol.

Notes
Hygroscopic. Store under inert gas. Protect from light. Store away from oxidizing materials.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text