Thermo Scientific Chemicals

Phenyltrimethoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: A13827.22
100 g, Each
Thermo Scientific Chemicals

Phenyltrimethoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: A13827.22
100 g, Each
Quantity
Catalog number: A13827.22
also known as A13827-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2996-92-1
IUPAC Name
trimethoxy(phenyl)silane
Molecular Formula
C9H14O3Si
InChI Key
ZNOCGWVLWPVKAO-UHFFFAOYSA-N
SMILES
CO[Si](OC)(OC)C1=CC=CC=C1
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Form
Liquid
Refractive Index
1.4700-1.4750 @ 20?C
Assay (GC)
≥96.0%

Description

Phenyltrimethoxysilane is used for nanoelectronics and material science research. It is used as crosslinking agent for silicone resin and to produce polymer organic silicon compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltrimethoxysilane is used for nanoelectronics and material science research. It is used as crosslinking agent for silicone resin and to produce polymer organic silicon compound.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Desiccate at room temperature. Store under inert gas. Incompatible with Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text