Thermo Scientific Chemicals

Dansyl chloride, 96%, Thermo Scientific Chemicals

Catalog number: A13828.06
5 g, Each
Thermo Scientific Chemicals

Dansyl chloride, 96%, Thermo Scientific Chemicals

Catalog number: A13828.06
5 g, Each
Quantity
Catalog number: A13828.06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
605-65-2
IUPAC Name
5-(dimethylamino)naphthalene-1-sulfonyl chloride
Molecular Formula
C12H12ClNO2S
InChI Key
XPDXVDYUQZHFPV-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=CC2=C(C=CC=C12)S(Cl)(=O)=O
Identification (FTIR)
Conforms
Form
Powder
Appearance (Color)
Yellow to orange
Assay (HPLC)
≥95.0%

Description

Dansyl chloride is a fluorogenic reagent used for protein, N-terminal amino acid and peptide detection by reverse phase high performance liquid chromatography (HPLC). It is also used to determine the degree of labeling in chymotrypsin and ovalbumin. It acts as a precursor for the synthesis of other derivatives like dansyl amide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dansyl chloride is a fluorogenic reagent used for protein, N-terminal amino acid and peptide detection by reverse phase high performance liquid chromatography (HPLC). It is also used to determine the degree of labeling in chymotrypsin and ovalbumin. It acts as a precursor for the synthesis of other derivatives like dansyl amide.

Solubility
Soluble in dimethyl fluoride, acetone, chloroform, pyridine, benzene and dioxane. Insoluble in water.

Notes
Store in cool place. Moisture sensitive. Incompatible with strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text