Thermo Scientific Chemicals

Salicylaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: A13833.0E
2500 g, Each
Thermo Scientific Chemicals

Salicylaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: A13833.0E
2500 g, Each
Quantity
Catalog number: A13833.0E
also known as A13833-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
90-02-8
IUPAC Name
2-hydroxybenzaldehyde
Molecular Formula
C7H6O2
InChI Key
SMQUZDBALVYZAC-UHFFFAOYSA-N
SMILES
OC1=CC=CC=C1C=O
Form
Liquid
Refractive Index
1.5715-1.5745 @ 20?C
Appearance (Color)
Clear, colourless to pale yellow
Assay (GC)
≥98.5%
Impurity content
Phenol: ≤1.0%

Description

Precursor to a variety of chelating agents.Salicylaldehyde is used as a precursor to prepare catechol, benzofuran, salicylaldehydimine and 3-carbethoxycoumarin. It is used for colorimetric assay of isoleucine as well as utilized for the determination of primary amine by fluorescence method. It is a key starting material to a various chelating agents. It is used as flavor and fragrance components. Its derivative salicylaldehyde thiosemicarbazone is used for the simultaneous determination of molybdenum and iron by spectrophotometric method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Precursor to a variety of chelating agents.Salicylaldehyde is used as a precursor to prepare catechol, benzofuran, salicylaldehydimine and 3-carbethoxycoumarin. It is used for colorimetric assay of isoleucine as well as utilized for the determination of primary amine by fluorescence method. It is a key starting material to a various chelating agents. It is used as flavor and fragrance components. Its derivative salicylaldehyde thiosemicarbazone is used for the simultaneous determination of molybdenum and iron by spectrophotometric method.

Solubility
Miscible with alcohol, ether, acetone, benzene and most organic solvents. Slightly miscible with water.

Notes
Incompatible with acids, strong reducing agents, strong oxidizing agents, strong bases and halogens.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text