N-Benzylaminoacetaldehyde diethyl acetal, 96%, Thermo Scientific Chemicals
N-Benzylaminoacetaldehyde diethyl acetal, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzylaminoacetaldehyde diethyl acetal, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A13841.14
also known as A13841-14
Price (USD)/ Each
399.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
399.00
Add to cart
N-Benzylaminoacetaldehyde diethyl acetal, 96%, Thermo Scientific Chemicals
Catalog numberA13841.14
Price (USD)/ Each
399.00
-
Add to cart
Chemical Identifiers
CAS61190-10-1
IUPAC Namebenzyl(2,2-diethoxyethyl)azanium
Molecular FormulaC13H22NO2
InChI KeySXFVQTYQHWRYOS-UHFFFAOYSA-O
SMILESCCOC(C[NH2+]CC1=CC=CC=C1)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive Index1.4865-1.4925 @ 20°C
FormLiquid
Assay (GC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only