Cyclopropylamine, 98+%, Thermo Scientific Chemicals
Cyclopropylamine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopropylamine, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A13844.09
also known as A13844-09
Price (USD)/ Each
43.50
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
43.50
Add to cart
Cyclopropylamine, 98+%, Thermo Scientific Chemicals
Catalog numberA13844.09
Price (USD)/ Each
43.50
-
Add to cart
Chemical Identifiers
CAS765-30-0
IUPAC Namecyclopropanamine
Molecular FormulaC3H7N
InChI KeyHTJDQJBWANPRPF-UHFFFAOYSA-N
SMILESNC1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Refractive Index1.4185-1.4225 @ 20?C
Cyclopropylamine) is used in the preparation of N-[4-(4-fluoro)phenyl-2-aminothiazol-5-yl]pyrimidin-2-yl-alkylamine derivatives. It is also used to prepare Pt(CPA)2(bismethylthiomethylenepropanedioate) and Pt(CPA)2(bisethylthiomethylenepropanedioate) complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropylamine) is used in the preparation of N-[4-(4-fluoro)phenyl-2-aminothiazol-5-yl]pyrimidin-2-yl-alkylamine derivatives. It is also used to prepare Pt(CPA)2(bismethylthiomethylenepropanedioate) and Pt(CPA)2(bisethylthiomethylenepropanedioate) complexes.

Solubility
Miscible with water, ethanol,ether and chloroform.

Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
RUO – Research Use Only

General References:

  1. Manakhov, A.; Nečas, D.; Čechal, J.; Pavliňák, D.; Eliáš, M.; Zajíčková, L. Deposition of stable amine coating onto polycaprolactone nanofibers by low pressure cyclopropylamine plasma polymerization. Thin Solid Films 2015, 581, 7-13.
  2. Miyamura, S.; Araki, M.; Suzuki, T.; Yamaguchi, J.; Itami, K. Stereodivergent Synthesis of Arylcyclopropylamines by Sequential C-H Borylation and Suzuki-Miyaura Coupling. Angew. Chem. Int. Ed. 2015, 54 (3), 846-851.