Thermo Scientific Chemicals

Cyclopropylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A13844.30
250 g, Each
Thermo Scientific Chemicals

Cyclopropylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A13844.30
250 g, Each
Quantity
Catalog number: A13844.30
also known as A13844-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
765-30-0
IUPAC Name
cyclopropanamine
Molecular Formula
C3H7N
InChI Key
HTJDQJBWANPRPF-UHFFFAOYSA-N
SMILES
NC1CC1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Refractive Index
1.4185-1.4225 @ 20?C

Description

Cyclopropylamine) is used in the preparation of N-[4-(4-fluoro)phenyl-2-aminothiazol-5-yl]pyrimidin-2-yl-alkylamine derivatives. It is also used to prepare Pt(CPA)2(bismethylthiomethylenepropanedioate) and Pt(CPA)2(bisethylthiomethylenepropanedioate) complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropylamine) is used in the preparation of N-[4-(4-fluoro)phenyl-2-aminothiazol-5-yl]pyrimidin-2-yl-alkylamine derivatives. It is also used to prepare Pt(CPA)2(bismethylthiomethylenepropanedioate) and Pt(CPA)2(bisethylthiomethylenepropanedioate) complexes.

Solubility
Miscible with water, ethanol,ether and chloroform.

Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text